Learn More
Thermo Scientific™ KingFisher™ Flex 96 Deep Well head
The KingFisher™ Flex 96 Deep Well head is a magnetic head for the KingFisher™ Flex Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 96 deep-well format system or a supplemental head for the 24 deep-well format system.
Brand:  Thermo Scientific™ 24074430
Product Code. 12515977
  • 4450.00 € / Each
Explore more special offers
Description
Description
The KingFisher™ Flex 24 Deep Well head is a magnetic head for the KingFisher™ Flex Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 96 deep-well format system or a supplemental head for the 24 deep-well format system. Reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.
The KingFisher™ Flex 24 Deep Well head is a magnetic head for the KingFisher™ Flex Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 96 deep-well format system or a supplemental head for the 24 deep-well format system. Reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KingFisher™
Each
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Flex 96 Deep Well head >