missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Untreated 245mm Square BioAssay Dishes
GREENER_CHOICE

Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.

Brand:  Corning™ 431272

 View more versions of this product

Product Code. 10002611

  • 231.96 € / Pack of 16
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Improve the efficiency of microbial assays including library replication, colony picking, and plaque lift studies.

 

  • Polystyrene dishes feature stacking beads for stable stacking
  • Compatible with various automated colony picking and gridding robots
  • Supplied with vented lid to provide consistent gas exchange
  • Sterile, certified nonpyrogenic
  • Patented (U.S. Patent No.. 5,856,176) design allows for easier handling

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
150 mL (Well)
500 sq. cm
Polystyrene
Sterile
Lid, Without Handles
245 x 245 x 26 mm
245 mm
18 mm
Closure Included
Square
Clear
Single-use
Square
Cell Culture Dish
245 x 245 mm
245 mm
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Untreated 245mm Square BioAssay Dishes > With handles; L x W x H: 245 x 245 x 26mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.