Learn More
Corning™ Untreated 245mm Square BioAssay Dishes

Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.
Brand: Corning™ 431301
Description
Improve the efficiency of microbial assays including library replication, colony picking, and plaque lift studies.
- Polystyrene dishes feature stacking beads for stable stacking
- Compatible with various automated colony picking and gridding robots
- Supplied with vented lid to provide consistent gas exchange
- Sterile, certified nonpyrogenic
- Patented (U.S. Patent No.. 5,856,176) design allows for easier handling

Specifications
Clear | |
Polystyrene | |
150 mL (Well) | |
500 sq. cm | |
Polystyrene | |
Sterile | |
Lid, Without Handles | |
245 x 245 x 20 mm | |
245 mm | |
12.5 mm |
Closure Included | |
Square | |
Clear | |
Single-use | |
Square | |
Cell Culture Dish | |
245 x 245 mm | |
245 mm | |
25 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.