missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Spheroid Microplate
BIOPHARMAGREENER_CHOICE

Treated with an ultra-low attachment surface to enable uniform and reproducible 3D multicellular spheroid formation. Corning™ 384-Well Spheroid Microplate permit direct microscopic viewing and readings for top and bottom reading instruments.

Brand:  Corning™ 3830

2428.62 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 15367850

  • 2595.00 € / Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.

  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment (ULA) surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate without the need for transfer to a new microplate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
0.06 cm2
Clear
20 to 80 μL
Ultra-low Binding
Minimizing protein absorption, enzyme activation and cellular activation
10/Pk.
With Lid
16 x 24
384
Sterile
Round Bottom (U-well)
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number32424016 Certificate TypeCertificate of Quality Date02/04/2025 Catalog Number3830
Lot Number28524039 Certificate TypeCertificate of Quality Date05/03/2025 Catalog Number3830
Lot Number26424008 Certificate TypeCertificate of Quality Date08/02/2025 Catalog Number3830
Lot Number25724004 Certificate TypeCertificate of Quality Date25/01/2025 Catalog Number3830
Lot Number24324022 Certificate TypeCertificate of Quality Date03/01/2025 Catalog Number3830
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 384-Well Spheroid Microplate > 10/Pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.