missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Clear Ultra Low Attachment Microplates
BIOPHARMAGREENER_CHOICE

Covalently bonded hydrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation. Corning™ 96-Well Clear Ultra Low Attachment Microplates allow for lentivirus transduction of human osteoclast precursor cells.

Brand:  Corning™ 7007

471.36 EUR valid until 2025-03-31
Use promo code "24073" to get your promotional price.


Product Code. 10023683

  • 650.00 € / Pack of 24
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ultra low attachment surface with covalently bonded hydrogel that minimizes cell attachment, protein absorption, enzyme activation and cellular activation
  • The surface is noncytotoxic, biologically inert and nondegradable
  • Round bottoms with 330μL total volume
  • Recommended working volumes of 75 to 200μL
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Individually wrapped
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
0.32 cm2
Ultra-Low Attachment
Polystyrene
75 to 200 μL
Ultra-low Binding
Lids
With Lid
8 x 12
Clear
96
Sterile
Round
24/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number32424042 Certificate TypeCertificate of Quality Date29/03/2025 Catalog Number7007
Lot Number32324025 Certificate TypeCertificate of Quality Date29/03/2025 Catalog Number7007
Lot Number32024021 Certificate TypeCertificate of Quality Date29/03/2025 Catalog Number7007
Lot Number31324044 Certificate TypeCertificate of Quality Date25/03/2025 Catalog Number7007
Lot Number31124022 Certificate TypeCertificate of Quality Date19/03/2025 Catalog Number7007
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 96-Well Clear Ultra Low Attachment Microplates > 96-well; 330μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.