Learn More
Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE
Enhanced signal-to-noise ratio
Brand:  Corning™ 3604
 View more versions of this product
Product Code. 10377171
  • 1572.07 € / Pack of 100
Estimated Shipment: 18-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Standard
    8 x 12
    Plate readers, liquid handlers and robotic systems
    96
    Non-sterile
    Flat Bottom (F-well)
    100/Cs.
    100/Cs.
    4°C to 37°C
    Without Lid
    White, Clear Bottom
    Polystyrene
    75 to 200 μL
    Non-binding
    For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    25/Pk.
    39°F to 99°F
    Clear
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Information
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates > White and Clear; Standard; No; NBS; Flat