missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates

Enhanced signal-to-noise ratio
1190.17 € - 1572.07 €
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
Material | Polystyrene |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
10377171
![]() |
Corning™
3604 |
White, Clear Bottom |
1572.07 €
Pack of 100 |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
10649541
![]() |
Corning™
3651 |
Black, Clear Bottom |
1190.17 €
Pack of 100 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
Description
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Non-binding | |
For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays | |
25/Pk. | |
39°F to 99°F | |
Clear |
Without Lid | |
Plate readers, liquid handlers and robotic systems | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. | |
4°C to 37°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates