missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.
Brand: Corning™ 3650
1044.18 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Standard | |
8 x 12 | |
Black | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. | |
4° to 37°C |
Without Lid | |
4 inner packs of 25 | |
Plate readers, liquid handlers and robotic systems | |
360 μL | |
75 to 200 μL | |
Non-binding | |
For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays | |
25/Pk. | |
39°F to 99°F | |
Black |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction