missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates

Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.
771.00 € - 1301.62 €
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
Material | Polystyrene |
Product Code | Brand | Color | Well Shape | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Well Shape | Price | Quantity & Availability | |||||
10409331
![]() |
Corning™
3600 |
White | Flat Bottom (F-well) |
1301.62 €
Pack of 100 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
10261692
![]() |
Corning™
3641 |
Clear | Flat Bottom (F-well) |
771.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10276091
![]() |
Corning™
3650 |
Black | Flat Bottom (F-well) |
1190.17 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10111822
![]() |
Corning™
3605 |
White | Round |
1064.77 €
Pack of 100 |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
Description
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Non-binding | |
100/Cs. | |
100/Cs. | |
4° to 37°C |
Without Lid | |
Plate readers, liquid handlers and robotic systems | |
96 | |
Non-sterile | |
For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays | |
25/Pk. | |
39°F to 99°F |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates