missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE

Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.

Brand:  Corning™ 3600

 View more versions of this product

Product Code. 10409331

  • 1301.62 € / Pack of 100
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Standard
    8 x 12
    Plate readers, liquid handlers and robotic systems
    360 μL
    75 to 200 μL
    Non-binding
    For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    25/Pk.
    39°F to 99°F
    White
    Without Lid
    White
    Polystyrene
    96
    Non-sterile
    Flat Bottom (F-well)
    100/Cs.
    100/Cs.
    4° to 37°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Product Information

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Corning™ 96-Well Nonbinding Surface (NBS™) Microplates > White; Standard; No; NBS; Flat

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.