missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Cell Strainers
BIOPHARMA

Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.

Brand:  Corning™ 431750

97.65 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 15360801

  • 105.00 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • Strong nylon mesh available in three pore sizes (40, 70 or 100 μm) for variety of applications
  • Fits all major brands of 50 mL conical tubes
  • Non cytotoxic device has color-coded polypropylene frames
  • Grip area for handling with forceps
  • Sterilized by gamma irradiation
  • Use for pre-filtering solutions containing particulates >40μm and for cleaning cell suspensions prior to sub culturing, counting and analysis or freezing
  • Individually packed in easy to open bags

 

For preparation of:

  • Single cell suspensions of blood cells
  • Specimens for primary cell cultures and immunogens
  • Primary cells from tissue

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nylon
Polypropylene
Conical Tubes, 50 mL
Blue
Sterile
40 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number266367 Certificate TypeCertificate of Quality Date11/03/2025 Catalog Number431750
Lot Number264724 Certificate TypeCertificate of Quality Date20/02/2025 Catalog Number431750
Lot Number264359 Certificate TypeCertificate of Quality Date23/01/2025 Catalog Number431750
Lot Number255622 Certificate TypeCertificate of Quality Date18/12/2024 Catalog Number431750
Lot Number249589 Certificate TypeCertificate of Quality Date17/09/2024 Catalog Number431750
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Sterile Cell Strainers > 40 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.