Learn More
Corning™ Sterile Cell Strainers

Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.
Brand: Corning™ 431751
99.75 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Strong nylon mesh available in three pore sizes (40, 70 or 100 μm) for variety of applications
- Fits all major brands of 50 mL conical tubes
- Non cytotoxic device has color-coded polypropylene frames
- Grip area for handling with forceps
- Sterilized by gamma irradiation
- Use for pre-filtering solutions containing particulates >40μm and for cleaning cell suspensions prior to sub culturing, counting and analysis or freezing
- Individually packed in easy to open bags
For preparation of:
- Single cell suspensions of blood cells
- Specimens for primary cell cultures and immunogens
- Primary cells from tissue

Specifications
Nylon | |
Polypropylene | |
Conical Tubes, 50 mL |
White | |
Sterile | |
70 μm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.