Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X

With low endotoxin content.
Brand: Corning™ 21-031-CV
58.14 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Specifications
No Calcium, No Magnesium | |
1 X | |
6 x 500 mL | |
7.3 to 7.5 |
2° to 30°C | |
DPBS | |
Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.