missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) without Glutamine
BIOPHARMA

Guaranteed low endotoxin levels ≤0.025EU/mL.

Brand:  Corning™ 17-305-CV

 View more versions of this product

Product Code. 15303571

  • 171.90 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.  This research indicated that additions made to BME aided cell propagation.  MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells.  When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayer, such as fibroblast.  This formulation contains sodium bicarbonate and does not contain L-glutamine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
Sterile
No Glutamine, No Phenol Red
7 to 7.4
MEM Eagle/EBSS
Earle's Balanced Salt Solution (EBSS)
6 x 500 mL
2° to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number15724006 Certificate TypeCertificate of Quality Date04/02/2025 Catalog Number17-305-CV
Lot Number03324007 Certificate TypeCertificate of Quality Date01/08/2024 Catalog Number17-305-CV
Lot Number21422015 Certificate TypeCertificate of Analysis Date08/03/2023 Catalog Number17-305-CV
Lot Number32121010 Certificate TypeCertificate of Analysis Date01/09/2022 Catalog Number17-305-CV
Lot Number24120005 Certificate TypeCertificate of Analysis Date09/07/2021 Catalog Number17-305-CV
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) without Glutamine > with Earle's Salts, without L-glutamine and Phenol Red; 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.