Learn More
Corning™ Trace Elements
BIOPHARMA
For Research applications
Brand:  Corning™ 25-023-CI
58.53 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 15353641
  • 66.13 € / 100mL
Explore more special offers
Description
Description
cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • Trace Elements A includes Cupric Sulfate, Ferric Citrate, Sodium Selenite, and Zinc Sulfate
  • Trace Elements B includes Ammonium Molybdate, Ammonium Vanadate, Manganese Sulfate, Nickel Sulfate, Sodium Silicate, Stannous Chloride and Hydrochloric Acid
  • Trace Elements C includes Aluminum Chloride, Barium Acetate, Cadmium Chloride, Chromic Chloride, Cobalt Dichloride, Germanium Dioxide, Potassium Bromide, Potassium Iodide, Rubidium Chloride, Silver Nitrate, Sodium Fluoride, and Zirconyl Chloride
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Liquid
Sterile-filtered
100 mL
Trace Elements C
Trace Element
Plastic Bottle