missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Tryptic Soy Broth, Animal-Free, Liquid
BIOPHARMA

For an accurate assessment of sterility throughout the manufacturing process

Brand:  Corning™ 46-060-CI

 View more versions of this product

Product Code. 15333591

  • 23.64 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Tryptic Soy Broth is a sterile-filtered, ready-to-use solution for use in sterility testing and validation in the manufacture of a liquid product. The use of this medium in place of the product intended for production allows for the assessment of the capability of an aseptic manufacturing process. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious micro-organisms. This growth medium has animal-free components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.

Order Info

Shipped under ambient conditions

TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP
Liquid
100 mL
15°C to 30°C
For testing the sterility of animal-free biopharmaceutical processes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number34524025 Certificate TypeCertificate of Quality Date03/04/2025 Catalog Number46-060-CI
Lot Number23924002 Certificate TypeCertificate of Quality Date01/01/2025 Catalog Number46-060-CI
Lot Number31023007 Certificate TypeCertificate of Quality Date04/09/2024 Catalog Number46-060-CI
Lot Number31023007 Certificate TypeCertificate of Analysis Date17/06/2024 Catalog Number46-060-CI
Lot Number20223002 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number46-060-CI
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Tryptic Soy Broth, Animal-Free, Liquid > 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.