Learn More
Corning™ Tryptic Soy Broth, Animal-Free, Liquid

For an accurate assessment of sterility throughout the manufacturing process
23.64 € - 1401.36 €
Specifications
Certifications/Compliance | USP |
---|---|
Content And Storage | 15°C to 30°C |
Form | Liquid |
For Use With (Application) | For testing the sterility of animal-free biopharmaceutical processes |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15333591
![]() |
Corning™
46-060-CI |
100 mL |
23.64 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15363721
![]() |
Corning™
46-060-LB |
20,000 mL |
1401.36 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptic Soy Broth is a sterile-filtered, ready-to-use solution for use in sterility testing and validation in the manufacture of a liquid product. The use of this medium in place of the product intended for production allows for the assessment of the capability of an aseptic manufacturing process. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious micro-organisms. This growth medium has animal-free components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.
Order Info
Shipped under ambient conditions
Specifications
USP | |
15°C to 30°C | |
Liquid | |
For testing the sterility of animal-free biopharmaceutical processes |
Safety and Handling
missing translation for 'shelfLife' : 1 Year
Your input is important to us. Please complete this form to provide feedback related to the content on this product.