Learn More
Corning™ Tryptic Soy Broth, Animal-Free, Liquid
BIOPHARMA
For an accurate assessment of sterility throughout the manufacturing process
Brand:  Corning™ 46-060-LB
 View more versions of this product
Product Code. 15363721
  • 1401.36 € / 20 L
Explore more special offers
Description
Description
Tryptic Soy Broth is a sterile-filtered, ready-to-use solution for use in sterility testing and validation in the manufacture of a liquid product. The use of this medium in place of the product intended for production allows for the assessment of the capability of an aseptic manufacturing process. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious micro-organisms. This growth medium has animal-free components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.
Order Info
Shipped under ambient conditions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP
Liquid
20,000 mL
15°C to 30°C
For testing the sterility of animal-free biopharmaceutical processes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Tryptic Soy Broth, Animal-Free, Liquid > 20 L