Learn More
Corning™ IPTG, Powder
BIOPHARMA
Produced under highest industry standards
Brand:  Corning™ 46-102-RF
Product Code. 15393721
  • 55.90 € / 1g
Explore more special offers
Description
Description
IPTG (isopropylthiogalactoside)
  • Inducer of lac operon in bacteria
  • Used with X-gal for detecting lac gene activity
  • For screening of clones containing gene of interest in lac operon
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For screening clones when used with X-Gal to detect lac gene activity
1000 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number33124023 Certificate TypeCertificate of Quality Date05/02/2025 Catalog Number46-102-RF
Lot Number46102052 Certificate TypeCertificate of Analysis Date22/08/2023 Catalog Number46-102-RF
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ IPTG, Powder > IPTG; 1g