Learn More
Quickfit™ Still Head Vertical Recovery Adapter
Used in distillation assemblies in which the condenser is assembled in the vertical mode
Brand: Quickfit SH2/22
Description
Used in distillation assemblies in which the condenser is assembled in the vertical mode
- Used in distillation assemblies in which the condenser is assembled in the vertical mode
Specifications
Used in distillation assemblies in which the condenser is assembled in the vertical mode | |
Borosilicate Glass | |
19/26 (Cone) |
Vertical Recovery Still Head | |
ISO 3585 |
Product Suggestions
Customers who viewed this item also viewed