missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Direct Overhead Drive 120mm Center Cap Assembly
For use with Corning Reusable spinner flasks
Brand: Corning™ 4519-134
This item is not returnable.
View return policy
Description

Specifications
8L and 15L Corning Reusable spinner flasks with 2.5 in. tubing | |
1/Pk. | |
Reusable |
1/Cs. | |
Center Cap Assembly | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Direct Overhead Drive 120mm Center Cap Assembly > For 8L and 15L flasks; 2-1/4 in. tubing
Spot an opportunity for improvement?Share a Content Correction