Learn More
Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)
GREENER_CHOICE
Enhance the proliferation of a wide variety of cell types
Brand:  Corning™ 354010
 View more versions of this product
Product Code. 11573520
  • 283.00 € / 100µg
Explore more special offers
Description
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.
Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Natural
A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research
100 μg
Male Mouse Submaxillary Glands
Growth Factor
Lyophilized from ammonium acetate, carrier free, reconstitute in dH2O
>98% by SDS-PAGE and pH 9.9 PAGE