Learn More
Corning™ Flexible Packaging Systems: MPC Connector Tubing
Clear tubing; female MPC connector
Brand:  Corning™ 91-700-04
 View more versions of this product
Product Code. 17243594
  • 53.15 € / Each
Explore more special offers
Description
Description
  • Chemically resistant
  • Heat sealable
  • Flexible
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Connector tubing
cGMPs, FDA Regulation 21 CFR part 820 and ISO 13485 Quality
Male luer Cap
Fits tubing 91.44 cm
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Flexible Packaging Systems: MPC Connector Tubing > 36 in.; Female MPC Connector, Female Luer, and Male Luer Cap