Learn More
Corning™ Frosted Microscope Slides
Made from water-white glass to maximize clarity. Easy to write on.
Brand: Corning™ 2949-75X25
This item is not returnable.
View return policy
Description
These Corning™ microscope slides are made from water-white glass to maximize clarity and are frosted at one end, both sides. Slides are 0.9 to 1.10 mm thick. Corning™ frosted slides are highly legible and easy to write on because they are sandblasted instead of etched. To minimize waste, they are inspected for chips and rough edges before being packaged. Products are packaged in 1/2 gross boxes (approx. 72 slides each). 10 gross = 20 boxes (1440 slides total)
Specifications
75 mm | |
1440 | |
One End, Both Sides | |
0.9 to 1.1 mm |
25 mm | |
72 | |
Soda Lime Glass |
COR019 CLS GL 035 DL Cover glass
File size: 236.4 kB
COR001 Lifescience Guid
File size: 7 MB
Product Content Correction
Product Title
Corning™ Frosted Microscope Slides > 1440/Cs.
Spot an opportunity for improvement?Share a Content Correction