Learn More
Invitrogen™ SYPRO™ Ruby Protein Blot Stain
Compatible with immunostaining (Western blotting), microsequencing and mass spectrometry
Brand: Invitrogen™ S11791
Description
SYPRO™ Ruby protein blot stain is a highly sensitive, ready-to-use fluorescent stain for proteins on nitrocellulose or PVDF membranes. Stained proteins can be viewed with a standard UV transilluminator or with a laser scanner.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature

Specifications
Store at room temperature and protect from light. | |
Fluorescence | |
Protein Blot Stain | |
200 mL | |
Protein |
In-Blot Detection | |
SYPRO Ruby | |
SYPRO™ | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.