Learn More
Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable
GREENER_CHOICE
Designed for universal fit with all common brands of pipet tips
Brand:  Corning™ 4072
 View more versions of this product
Product Code. 11317431
  • 222.15 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
Services
3 year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±4 to ±0.8%
200μL Tip
200 μL Tip
Lambda Plus
2 to 20 μL
Key is included with each pipette for easy in-house calibration
3 Years
ISO8655
Autoclavable
1
Single-Channel Pipettor
200μL Tip
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable > 2-20 μL; 1; None