Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase (200 U/μL)
Thermo Scientific Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT.
Brand: Thermo Scientific™ EP0753
727.13 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Thermo Scientific Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT. The engineered enzyme features dramatically improved thermostability, 50X higher processivity, robustness, and increased synthesis rate compared to wild type M-MuLV RT.
The eliminated RNase H activity enables the enzyme to produce very long RNA transcripts up to 20 kb. Due to its high thermostability, the enzyme maintains full activity during the entire reverse transcription reaction and generates high yields of cDNA. The reaction temperature can be increased to 65°C for efficient transcription of RNA regions with a high secondary structure or to improve specificity using gene specific primers. The extremely high processivity of Maxima H Minus enzyme results in increased resistance to common reaction inhibitors, such as guanidine, formamide, and ethanol.
Features of Maxima H Minus Reverse Transcriptase include:
• Thermostable—90% active after incubation at 50°C for 60 minutes in a reaction mixture
• Active up to 65°C
• High yields of full-length cDNA up to 20 kb
• High sensitivity—reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
• Efficient—complete cDNA synthesis in 15 to 30 minutes
• Increased resistance to common reaction inhibitors
• Incorporates modified nucleotides
Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Reverse transcription at elevated temperatures to reduce effects of secondary structure
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• Analysis of RNA by primer extension
Related products
• Maxima H Minus Reverse Transcriptase (Cat. No. EP0752)
• Maxima H Minus First Strand cDNA Synthesis Kit (Cat. No. K1652)
• Maxima H Minus First Strand cDNA Synthesis Kit, with dsDNase (Cat. No. K1681)
• Maxima H Minus cDNA Synthesis Master Mix (Cat. No. M1661)
• Maxima H Minus cDNA Synthesis Master Mix, with dsDNase (Cat. No. M1681)
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

Specifications
• Maxima H Minus Reverse Transcriptase, 4 x 10,000 units (200 U/μL) |
|
High | |
Reverse Transcription | |
4 x 10,000 Units | |
Reduced | |
200 U/μL | |
Separate components | |
Up to 20 kb |
Stand-alone Enzyme | |
15 to 30 min. | |
50°C to 55°C | |
Maxima H Minus | |
Dry Ice | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.