Learn More
Corning™ Reusable Gas Venting Cap for Vertical Sidearm Flasks
Used to provide gases into larger spinner flasks with vertical sidearms during cell culture
Brand:  Corning™ 4519-177
Product Code. 10042132
  • 375.49 € / Each
Estimated Shipment: 17-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used to provide gases into larger spinner flasks with vertical sidearms.
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • Gas filters are PTFE, 0.2 μm porosity.
  • The 316 stainless steel tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vertical sidearm flasks
1/Pk.
Non-sterile
1/Cs.
Fittings
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Reusable Gas Venting Cap for Vertical Sidearm Flasks > For Use With: Vertical sidearm flasks, Fittings