missing translation for 'onlineSavingsMsg'
Learn More

Corning™ SOC Medium
BIOPHARMA

Maximizes transformation efficiency of E. coli competent cells

Brand:  Corning™ 46-003-CR

Product Code. 15323711

  • 119.41 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Store at Room Temperature.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Use to maximize transformation efficiency of competent E. coli cells
2% Tryptone, 0.5% Yeast Extract, 0.4% Glucose, 10 mm NaCl, 2.5 mm KCl, 5 mm mgCl2, 5 mm mgSO4
10 x 10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number23324015 Certificate TypeCertificate of Quality Date25/02/2025 Catalog Number46-003-CR
Lot Number16224010 Certificate TypeCertificate of Quality Date11/12/2024 Catalog Number46-003-CR
Lot Number24823021 Certificate TypeCertificate of Quality Date03/07/2024 Catalog Number46-003-CR
Lot Number24823022 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number46-003-CR
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ SOC Medium > SOC Medium, 10/pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.