missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Supporting Plates for Vortex Mixer

Securely holds test tubes and vials. Supporting Plates are compatible with Vortex Mixers.

Brand:  Fisherbrand™ A00000016

17.77 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 11702593

  • 19.55 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Soft foam or rubber to prevent breakage
  • Black in color
  • Availalbe in different diameters
Specifications

Specifications

Supporting Plate
50 mm
CLASSIC, ZX3, WIZARD, ZX4, TX4, RX3
Rubber
Supporting Plate, 50mm dia.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Supporting Plates for Vortex Mixer > Outer Diameter: 50 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.