missing translation for 'onlineSavingsMsg'
Learn More
Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap Product Code.: 10692434
Add to basket

Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap
BIOPHARMA

Product Code. 10692434 Shop All Corning Products
150 mL
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
1 L
This item is not returnable. View return policy

Product Code. 10692434

Brand: Corning™ 431430

Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Linerless, one-piece, autoclavable orange GL45 threaded polypropylene plug-seal caps

Space-Saving square shape requires less space (13 to 20%) on the shelf or in the autoclave. Corning™ Square Polycarbonate Storage Bottles are ideal for mixing, sampling and storage.

Corning™ square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.


Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

  • Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.

  • Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

  • Screened white enamel graduations are easier to see than molded graduations

  • Sterilized by gamma radiation

  • Large white marking spot for easier identification

  • Bottles can be autoclaved once at 121oC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.

  • Optional reusable caps with silicone septa (Corning Cat. Nos. 1395-45HTSC, 1395-45SS, 1395-45TS) for syringe sampling or introduction of reagents are available for these caps.

  • Note: Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity 4.5 oz., 150 mL
    Capacity (Metric) 150 mL
    Capacity (English) 4.5 oz.
    Material Polycarbonate
    No. per Case 24/Cs.
    No. per Pack 12/Pk.
    Packaging Individually wrapped clear plastic
    Sterility Sterile
    Closure Color Orange
    Graduations 50 to 150 mL
    Autoclavable Autoclavable
    Shape Square
    Closure Material Polypropylene
    Closure Size GL 45
    Closure Type Screw Cap
    Show More Show Less

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number15924011 Certificate TypeCertificate of Quality Date27/04/2025 Catalog Number431430
    Lot Number10324006 Certificate TypeCertificate of Quality Date25/02/2025 Catalog Number431430
    Lot Number30223013 Certificate TypeCertificate of Quality Date06/09/2024 Catalog Number431430
    Lot Number01024001 Certificate TypeCertificate of Quality Date15/05/2024 Catalog Number431430
    Lot Number30223013 Certificate TypeCertificate of Compliance Date19/04/2024 Catalog Number431430
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.