Learn More
Corning™ Storage Mat III
Fits Corning™ 96 round well plates and blocks. Corning™ Storage Mat III can be applied manually or with a storage mat applicator.
Brand:  Corning™ 3080
295.03 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 10428571
  • 379.25 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
(This is the suggested replacement for 3092 and 3094)
  • Fits Corning 96 round well plates and blocks
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • DNase- and RNase-free
  • Can be applied manually or with our Storage Mat Applicator
  • Nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNase and RNase-free
Non-sterile
25/Pk.
Round
EVA, Polymer
100/Cs.
Storage Mat III
96-well Plate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Storage Mat III > Round