Learn More
Invitrogen™ E-Gel™ Power Snap Camera
Camera designed for quick and effortless E-Gel™ agarose gel image capture and documentation when used with E-Gel™ Power Snap Electrophoresis Device.
Brand: Invitrogen™ G8200
Includes: E-Gel Power Snap Camera
Description
The E-Gel Power Snap Camera is designed for quick and effortless E-Gel agarose gel image capture and documentation when used with E-Gel Power Snap Electrophoresis Device (G8100)*. The cable-free, easy-to-dock E-Gel Power Snap Camera seamlessly integrates into the device, enabling instant image capture during the run without interruption of the workflow.
The E-Gel Power Snap Camera is optimized for use with E-Gel EX, E-Gel SYBR Safe, E-Gel Go!, E-Gel CloneWell II, and E-Gel SizeSelect II gels, and is not compatible with ethidium bromide stained gels.
Features of the E-Gel Power Snap Camera include:
- Capture high-resolution E-Gel agarose gel images instantly
- Adjust gel images with easy-to-use onboard software
- Store months of gel image files using internal camera memory
- Complete walk-away workflow with auto-capture feature
Instant capture and seamless integration
Capture high-resolution E-Gel agarose gel images on the go with the easy-to-use E Gel Power Snap Camera. The camera is directly docked to the E-Gel Power Snap Electrophoresis Device, requiring no external power supply or connection to a desktop computer. This enables the freedom to capture images instantly, on the spot, without leaving the bench.
Effortless use
This high-resolution digital color camera automatically captures E-Gel agarose gel images at optimal exposure settings without the need for focusing or image/gel adjustments. The auto-exposed image can be fine-tuned manually if needed.
Made for convenience
The integrated E-Gel Power Snap Camera touchscreen and onboard software provide an intuitive interface for quick gel image capture and editing. Store images using the Power Snap internal memory storage through months of camera use or easily transfer the images to an external USB memory device.
Freedom to walk away
The E-Gel Power Snap Camera features an automatic gel capture workflow. This enables the capture of multiple images at various time intervals after you have walked away. Find all of your images stored in internal image gallery when you return.
*The E-Gel Power Snap Camera is designed only for E-Gel agarose gel imaging and functions only when used with E-Gel Power Snap Electrophoresis Device.
Services
1 year warranty
Specifications
E-Gel Camera | |
1 camera | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2848124100076 | Certificate of Compliance | 13/02/2025 | G8200 |
2848125010007 | Certificate of Compliance | 12/02/2025 | G8200 |
2848124070012 | Certificate of Compliance | 12/02/2025 | G8200 |
2848124040018 | Certificate of Compliance | 12/02/2025 | G8200 |
2848124030119 | Certificate of Compliance | 12/02/2025 | G8200 |