missing translation for 'onlineSavingsMsg'
Learn More

Corning™ TC-Treated Culture Dishes
GREENER_CHOICE

Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.

Brand:  Corning™ 431110

228.95 EUR valid until 2025-03-31
Use promo code "24073" to get your promotional price.


 View more versions of this product

Product Code. 10350602

  • 250.00 € / Pack of 16
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
  • Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
  • 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
  • 245 mm square dishes offer 500 cm2 growth surface
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Manufactured from optically clear virgin polystyrene
  • Sterile
  • Nonpyrogenic


Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.

Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.

Order Info

Package quantities differ. Check ordering table for details.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
Flat
Clear
Single-use
Polystyrene
Sterile
Cell Culture Petri Dish
Lid
4/Pk.
245 x 245 mm
245 mm
Yes
245 L x 245 W x 25 mm H
150 mL (Well)
500 cm2
Not Gridded
Square
Cell Culture
100 to 150 mL
16/Cs.
245 x 245 mm
245 mm
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.