Learn More
Corning™ Universal Fit Pipet Tips: Bulk Packed
GREENER_CHOICE
Designed to assure a reliable fit with major brand pipetters
Brand:  Corning™ 4862
 View more versions of this product
Product Code. 10005451
  • 39.66 € / Pack of 1000
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed to assure a reliable fit with major brand pipetters
  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 200 μL
Natural
Beveled, Round
Yes
Non-sterile
Bulk
Universal Fit
Polypropylene