missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ SARS-CoV-2 Proteome Microarray

SARS-CoV-2 Proteome Microarray with 4,883 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening, or biomarker discovery.

Brand:  PEPperCHIP™ SPC.051.001

Product Code. 16544780

  • 593.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Microarray Content: Full proteome of Whole proteome of Wuhan seafood market pneumonia virus isolate Wuhan-Hu-1 (GenBank ID: MN908947.3) as overlapping linear peptides
  • Peptide Length/Overlap: 15 aa / 13 aa
  • Number of Peptides/Spots: 4,883 peptides printed in duplicate and HA and polio control peptides
  • Related Products: PEPperCHIP Incubation Tray 3/1, PEPperCHIP anti-HA control antibodies, PepSlide Analyzer
Specifications

Specifications

PEGMA/PMMA
1 array copy per microarray
1 mm
Transparent
Microscopic Glass
Antibody validation, epitope mapping, serum screening, biomarker discovery
75.4 x 25 mm
Research Use Only
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
Individual barcode for each microarray
High-density Peptide Microarray
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PEPperCHIP™ SARS-CoV-2 Proteome Microarray > Size: 75.4 x 25 x 1 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.