Learn More
DuPont™ Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ Taped Seam Hooded Coveralls are composed of flash spun high density polyethylene.
Brand: DuPont™ D13395272
232.96 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Self-adhesive chin strap for tight seal of suit to face mask
- Stitched and over-taped seams for protection and strength
- Tyvek zipper and zipper flap for enhanced protection
- Elasticated face, wrists and ankles as well as glued-in elastic waist
- Elasticated thumb loops prevent sleeves from riding up
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as medical applications and when exposed to biological hazards
Order Info
dimSize

Specifications
Hooded Coveralls | |
HDPE | |
Elastic | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Chemicals, Liquid | |
116 to 124 cm |
2X-Large | |
White | |
Elastic | |
Hooded | |
Zipper with Self-adhesive Flap and Chin Flap | |
Category III, Type 4-B, 5-B and 6-B | |
Category III, Type 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), (EN 1149-5) | |
Maintenance and Dismantling Jobs in the Nuclear Industry, Pharmaceutical Manufacturing or in Research and Biosecurity Laboratories |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.