missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab COSA CIP 92 Liquid Detergent
COSA CIP 92 is a liquid, alkaline, surfactant-containing detergent suitable for the removal of organic soil like emulsions, strong fatty formulations and gel in for the pharmaceutical and cosmetic industries.
Brand: Ecolab 2319850
This item is not returnable.
View return policy
Description
- Suitable for CIP-systems
- Excellent removal of organic soil
- High soil-suspending power
- For general CIP application (Homogenizers, pipelines, tanks, etc): Concentration: 1 to 3%, Temperature: 45°C to 95°C (circulating manner), Contact time: depending on degree and kind of soiling.
- Before switching on the spray devices, it is recommended to heat up the solution to >50°C to prevent the formation of foam.
- Concentration, temperature and cleaning time can be optimized by evaluation of respective cleaning trials.
- Final rinse with water of minimum drinking water quality, ensuring all soil and product residues are removed.
Specifications
Alkaline Detergent | |
Concentrated Liquid | |
Unscented | |
Clear Yellow | |
Yellow | |
Filtered | |
22 kg |
No | |
11.5 to 12.5 | |
Non-sterile | |
Not Autoclavable | |
Single-use | |
No |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COSA CIP 92
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Alcohol ethoxylate,octanoic acid, sodium salt, Alkylamine ethoxylates, sodium hydroxide
SDS_17899864_EN.pdf
File size: 423.3 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ecolab COSA CIP 92 Liquid Detergent > pH: 11.5 to 12.5
Spot an opportunity for improvement?Share a Content Correction