missing translation for 'onlineSavingsMsg'
Learn More

D-Lysine, Thermo Scientific™

Brand:  Thermo Scientific Acros 469950010

74.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17720719

  • 74.67 € / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

D-Lysine
923-27-3
Beige to White
C6 H14 N2 O2
00008234
(R)-2,6-Diaminohexanoic acid
LYSINE
146.19
Crystalline powder
No special requirements
218°C
311°C
Conforms
Glass Bottle
-12° to -16°
≤5 %
(20°C, 589 nm) (c=1 aq. soln.)
1 g
Life science
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Lysine, Thermo Scientific™ > 1 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.