missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Assay Blocks

Manufactured from clarified polypropylene for optical clarity. Corning™ 96-Well Assay Blocks are ideal for compound storage, assay, and cell-growth applications.
206.00 € - 1175.00 €
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Binding Type | Not Treated |
Color | Clear |
Product Code | Brand | Sterility | Volume (Metric) Well | Well Shape | No. of Wells | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Volume (Metric) Well | Well Shape | No. of Wells | Price | Quantity & Availability | |||
10554522
![]() |
Corning™
3961 |
Non-sterile | 2000 μL | Conical Bottom (V-well) | 96 |
1175.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10779922
![]() |
Corning™
3957 |
Non-sterile | 500 μL | Round, Conical Bottom (V-well) | 96 |
799.57 €
Pack of 100 |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||
10689141
|
Corning™
3959 |
Non-sterile | 1000 μL | Round | 96 |
687.45 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10708212
![]() |
Corning™
3960 |
Sterile | 2000 μL | Conical Bottom (V-well) | 96 |
342.00 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||
10196761
![]() |
Corning™
3958 |
Sterile | 1000 μL | Round | 96 |
206.00 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Certifications
Certified DNase- and RNase-free
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
For compound storage, assay and cell-growth application |
Without Lid | |
Not Treated | |
Polypropylene | |
Non-treated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Assay Blocks