missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Assay Blocks
BIOPHARMA

Manufactured from clarified polypropylene for optical clarity. Corning™ 96-Well Assay Blocks are ideal for compound storage, assay, and cell-growth applications.

Brand:  Corning™ 3961

787.04 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 10554522

  • 1175.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

Certifications

Certified DNase- and RNase-free

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Not Treated
96-well Assay Blocks
2000 μL
Non-sterile
Conical Bottom (V-well)
100/Cs.
100/Cs.
Without Lid
20 inner packs of 5
Clear
Polypropylene
96
Non-treated
For compound storage, assay and cell-growth application
5/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number21624000 Certificate TypeCertificate of Quality Date01/04/2025 Catalog Number3961
Lot Number20124000 Certificate TypeCertificate of Quality Date22/02/2025 Catalog Number3961
Lot Number17824000 Certificate TypeCertificate of Quality Date01/02/2025 Catalog Number3961
Lot Number17724000 Certificate TypeCertificate of Quality Date03/12/2024 Catalog Number3961
Lot Number15824000 Certificate TypeCertificate of Quality Date15/10/2024 Catalog Number3961
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 96-Well Assay Blocks > V-bottom; No

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.