missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates

Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.
1005.64 € - 1040.00 €
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Black, Clear Bottom |
Material | Polystyrene |
Product Code | Brand | Color | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Surface Treatment | Price | Quantity & Availability | |||||
10082471
![]() |
Corning™
3631 |
Black, Clear Bottom | Non-treated |
1040.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10123811
![]() |
Corning™
3601 |
Black, Clear Bottom | High Binding |
1005.64 €
Pack of 100 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
Description
- Molded polystyrene with opaque walls and optically clear bottoms
- Bottoms are 60% thinner than conventional polystyrene microplates for lower background flourecence and enabling readings down to 340nm
- Opaque black or white walls prevent well to well crosstalk
- Recommended working volumes of 75 to 200μL
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
Order Info
Cat. No. 07-200-568 also available in a pack of 25.
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Flat Bottom (F-well) | |
100/Cs. |
Without Lid | |
Black, Clear Bottom | |
96 | |
Non-sterile | |
100 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates