missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates
GREENER_CHOICE

Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.

Brand:  Corning™ 3631

896.26 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 10082471

  • 1040.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Molded polystyrene with opaque walls and optically clear bottoms
  • Bottoms are 60% thinner than conventional polystyrene microplates for lower background flourecence and enabling readings down to 340nm
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 75 to 200μL
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards

Order Info

Cat. No. 07-200-568 also available in a pack of 25.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Black, Clear Bottom
Polystyrene
96
Non-sterile
Flat Bottom (F-well)
100
Without Lid
4 inner packs of 25
EIA/RIA
360 μL
75 to 200 μL
Non-treated
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
100/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates > Untreated; Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.