missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Microplate Aluminum Sealing Tape
GREENER_CHOICE

Provides tight seal to minimize evaporation and condensation. Corning™ 384 Well Microplate Aluminum Sealing Tape enables oil-free operation when used with thermocyclers with heated lids.

75.90 € - 102.55 €

Specifications

Material Aluminum
Sterility Non-sterile
No. per Case 100/Cs.
No. per Pack 100/Pk.
For Use With (Application) Easy application and removal for short and long-term storage, provide tight seal to minimize evaporation and condensation and enables oil-free operation when used with thermo cyclers with heated lids
View More Specs

Products 2
Product Code Brand Plate Compatibility Price Quantity & Availability  
Product Code Brand Plate Compatibility Price Quantity & Availability  
10629141
GREENER_CHOICE
View Documents
Corning™
6570
96-well Plate
75.90 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10651272
GREENER_CHOICE
View Documents
Corning™
6569
384-well Plate
102.55 €
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

  • Easy application and removal for short- and long-term storage
  • Provides tight seal to minimize evaporation and condensation
  • Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
  • Breathable sealing tape (Cat. No. 3345) allows gas exchange across the surface.
  • Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
Specifications

Specifications

Aluminum
100/Cs.
Easy application and removal for short and long-term storage, provide tight seal to minimize evaporation and condensation and enables oil-free operation when used with thermo cyclers with heated lids
Yes
Non-sterile
100/Pk.
Sealing Tape
-80°C to +150°C
Videos
SDS
Documents

Documents

Product Certifications