Learn More
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
Brand:  Corning™ 8161
161.36 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
 View more versions of this product
Product Code. 10104101
  • 189.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cellulose Acetate
Non-sterile
0.5 mL
0.22 μm
Spin-X
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane > Cellulose Acetate; 0.22 μm; No