Learn More
Corning™ Clear Polystyrene 96-Well Microplates
GREENER_CHOICE
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
191.00 € - 378.37 €
Specifications
Format Standard
Array 8 x 12
Binding Type Not Treated
Color Clear
Material Polystyrene
View More Specs
Products 8
Product Code Brand Well Shape Surface Treatment Sterility Lid Price Quantity & Availability  
Product Code Brand Well Shape Surface Treatment Sterility Lid Price Quantity & Availability  
10265362
GREENER_CHOICE
View Documents
Corning™
3788
Round Non-treated Sterile With Lid
340.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10367841
GREENER_CHOICE
View Documents
Corning™
3896
Conical Bottom (V-well) Non-treated Sterile Without Lid This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10035351
GREENER_CHOICE
View Documents
Corning™
3798
Round Non-treated Non-sterile Without Lid
293.57 €
Pack of 100
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
10631462
GREENER_CHOICE
View Documents
Corning™
3898
Conical Bottom (V-well) Treated Non-sterile Without Lid
378.37 €
Pack of 100
Estimated Shipment: 21-08-2025
to see stock.
Add to basket
10296381
GREENER_CHOICE
View Documents
Corning™
3367
Round Non-treated Sterile Without Lid
191.00 €
Pack of 50
In Stock
to see stock.
Add to basket
10163711
GREENER_CHOICE
View Documents
Corning™
3795
Round Non-treated Sterile Without Lid
259.33 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10766482
GREENER_CHOICE
View Documents
Corning™
3897
Conical Bottom (V-well) Non-treated Non-sterile Without Lid
310.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10216341
GREENER_CHOICE
View Documents
Corning™
3370
Flat Bottom (F-well) Non-treated Sterile With Lid
315.12 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
Standard
Not Treated
Polystyrene
75 to 200 μL
8 x 12
Clear
96
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Corning™ Clear Polystyrene 96-Well Microplates