missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
120.64 € - 246.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Color | Translucent |
Volume (Metric) | 1 mL |
For Use With (Equipment) | Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges |
Product Code | Brand | Product Type | Format | Quantity | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Format | Quantity | Sterility | Price | Quantity & Availability | |||
11763994
|
Corning™
4412 |
8-Strip Microplate Cluster Tube | Strip | 12 Strips/Rack, 120 Strips/Cs. | Non-sterile |
231.00 €
Pack of 120 |
In Stock
Log in to see stock. |
|||
10681432
|
Corning™
4413 |
8-Strip Microplate Cluster Tube | Strip | 12/Rack | Sterile |
242.70 €
Pack of 120 |
In Stock
Log in to see stock. |
|||
10256531
|
Corning™
4401 |
Microplate Cluster Tube | Tube | Bulk | Non-sterile |
120.64 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10085431
|
Corning™
4408 |
8-Strip Microplate Cluster Tube | Strip | Bulk, 120/Bag | Non-sterile |
146.72 €
Pack of 120 |
In Stock
Log in to see stock. |
|||
10285471
|
Corning™
4410 |
Microplate Cluster Tube | Tube | 96/Rack | Non-sterile |
236.06 €
Pack of 960 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||
10690112
|
Corning™
4411 |
Microplate Cluster Tube | Tube | 96/Rack | Sterile |
246.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.
Specifications
Autoclavable | |
Translucent | |
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
96 |
Polypropylene | |
1 mL | |
0.033 oz. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Cluster Tube System