missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Disposable Aspirating Pipets
BIOPHARMAGREENER_CHOICE

Constructed of polystyrene for strength, clarity, and durability. Corning™ Costar™ Disposable Aspirating Pipets are ideal for aseptically aspirating liquids by using vacuum suction.

Brand:  Corning™ 9186

 View more versions of this product

Product Code. 10332282

  • 596.68 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • One piece polystyrene construction
  • Sterile and certified nonpyrogenic
  • No graduations; unplugged
  • Tip shape: round

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aspirating
10.9 in.
Individual Clear Plastic Wrap
2 mL
Single-use
Polystyrene
Round
0.067 oz.
27.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers