Learn More
Corning™ Costar™ Snap Cap Microcentrifuge Tubes
Features a snap cap with a positive seal that allows for repeated opening and closing with flat-cap surfaces for convenient labeling. Corning™ Costar™ Snap Cap Microcentrifuge Tubes are compatible with leading microcentrifuge rotors.
Brand:  Corning™ 3621
 View more versions of this product
Product Code. 10439521
  • 365.92 € / Pack of 5000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Supplied nonsterile and are autoclavable.
  • External graduations and frosted writing spot for easy sample identification.
  • Positive seal design allows repeated opening and closing.
  • Flat cap surface for convenient labeling.
  • Withstands a maximum RCF of 17,000 x g.
  • RNase-/DNase-free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Snap Cap
Natural
Autoclavable
Natural
500/Pk.
Non-sterile
Snap Cap
17,000 x g
RNase, DNase-free
Polypropylene
1.7 mL
Polypropylene
5000/Pk.
Yes
Conical
Microcentrifuge Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Costar™ Snap Cap Microcentrifuge Tubes > 1.7 mL; Natural; 5000/Cs