Learn More
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile
GREENER_CHOICE
Specially designed Netwell™ carriers with handles allow simultaneous processing of up to 12 samples per carrier
261.90 €
Specifications
Type Tray
No. per Pack 25/Pk.
No. per Case 200/Cs.
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10621272
GREENER_CHOICE
View Documents
Corning™
3517
Black
261.90 €
Pack of 200
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
10357841
GREENER_CHOICE
View Documents
Corning™
3519
White
261.90 €
Pack of 200
In Stock
to see stock.
Add to basket
Description
Description
  • Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
  • Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
  • Each carrier kit contains eight carriers and eight handles
Specifications
Specifications
Tray
25/Pk.
200/Cs.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile